Anti-SerpinB8 Rabbit Polyclonal Antibody
Supplier: Sino Biological
Certificates
About this item
Produced in rabbits immunized with purified, recombinant Mouse SerpinB8 (rM SerpinB8; Catalog#50215-M08B; NP_035589.1; Met 1-Pro 374). SerpinB8 specific IgG was purified by Mouse SerpinB8 affinity chromatography.
Type: Primary
Antigen: SERPINB8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 103657-412
- Antigen Symbol:
- SerpinB8
- Antigen Name:
- Serpin family B member 8
- Antigen Synonyms:
- NK10|ovalbumin|Spi8|CAP-2|CAP2
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Mouse
- Host:
- Rabbit
- Isotype:
- IgG
- Western Blot:
- Yes
- ELISA:
- Yes
- ImmunoPrecipitation:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient