Anti-LXN Rabbit Monoclonal Antibody [clone:040]
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Latexin / LXN (rM Latexin / LXN; Catalog#50050-M07E; NP_058033.2; Glu 2-Glu 222).
Type: Primary
Antigen: LXN
Clonality: Monoclonal
Clone: 040
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
- Catalog No:
- 103656-366
- Antigen Symbol:
- LXN
- Antigen Name:
- Latexin
- Antigen Synonyms:
- ECI|TCI
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 040
- Reactivity:
- Mouse
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient