Anti-EBOV-VP24 Mouse Monoclonal Antibody [clone:08]
Supplier: Sino Biological
Certificates
About this item
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Ebola virus EBOV (subtype Bundibugyo, strain Uganda 2007) VP24 ( Catalog#40456-V07E; YP_003815439.1; Met1-Lys233). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Type: Primary
Antigen: VP24 Protein
Clonality: Monoclonal
Clone: 08
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG2b
Reactivity: Ebola virus EBOV (subtype Bundibugyo;strain Uganda 2007)
Specifications
- Catalog No:
- 103656-060
- Antigen Symbol:
- EBOV-VP24
- Antigen Name:
- EBOV virus matrix 24 protein
- Antigen Synonyms:
- VP24
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 08
- Reactivity:
- Ebola virus eBOV (subtype bundibugyo strain uganda 2007)
- Host:
- Mouse
- Isotype:
- IgG2b
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Specifications
Frequently Bought Together