Anti-EBOV-G Rabbit Polyclonal Antibody
Supplier: Sino Biological
Certificates
About this item
Produced in rabbits immunized with purified, recombinant Ebola virus EBOV (subtype Bundibugyo, strain Uganda 2007) GP1 / Glycoprotein ( Catalog#40368-V08B; ACI28624.1; Met1-Arg501). Ebola virus EBOV (subtype Bundibugyo, strain Uganda 2007) GP1 / Glycoprotein specific IgG was purified by Ebola virus EBOV (subtype Bundibugyo, strain Uganda 2007) GP1 / Glycoprotein affinity chromatography.
Type: Primary
Antigen: GP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Ebola virus EBOV (subtype Bundibugyo;strain Uganda 2007)
Specifications
- Catalog No:
- 103612-488
- Antigen Symbol:
- EBOV-G
- Antigen Name:
- Ebola virus Glycoprotein
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Ebola virus eBOV (subtype bundibugyo strain uganda 2007)
- Host:
- Rabbit
- Isotype:
- IgG
- Western Blot:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Specifications
Frequently Bought Together