Anti-NP-CoV Rabbit Polyclonal Antibody
Supplier: Sino Biological
Certificates
About this item
Produced in rabbits immunized with purified, recombinant MERS-CoV (NCoV / Novel coronavirus) Nucleoprotein / NP (Catalog#40068-V08B; AFS88943.1; Met1-Asp413). Total IgG was purified by Protein A affinity chromatography.
Type: Primary
Antigen: NP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: MERS-CoV
Specifications
- Catalog No:
- 103655-860
- Antigen Symbol:
- NP-CoV
- Antigen Name:
- Coronavirus Nucleocapsid
- Antigen Synonyms:
- Nucleoprotein|coronavirus Nucleocapsid|cov np|ncov NP|coronavirus NP|Nucleocapsid|NP|coronavirus Nucleoprotein|novel coronavirus Nucleoprotein
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- MERS-CoV
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
- Tested Applications:
- 8
Specifications
Frequently Bought Together