Anti-DBI Rabbit Polyclonal Antibody
Supplier: Sino Biological
Certificates
About this item
Produced in rabbits immunized with purified, recombinant Human DBI / ACBD1 (rh DBI / ACBD1; Catalog#15211-H07E; P07108-2; Met1-Ala87). DBI / ACBD1 specific IgG was purified by Human DBI / ACBD1 affinity chromatography.
Type: Primary
Antigen: DBI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Specifications
- Catalog No:
- 103611-626
- Antigen Symbol:
- DBI
- Antigen Name:
- Diazepam binding inhibitor, acyl-CoA binding protein
- Antigen Synonyms:
- CCK-RP|ACBD1|ACBP
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient