About this item
This antibody was obtained from a mouse immunized with purified, recombinant Human MOB1B / MOBKL1A (rh MOB1B / MOBKL1A; Catalog#14016-H09E; Q7L9L4; Met1-Arg216). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Type: Primary
Antigen: MOB4A
Clonality: Monoclonal
Clone: 01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG2a
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 103654-652
- Antigen Symbol:
- MOB1B
- Antigen Name:
- MOB kinase activator 1B
- Antigen Synonyms:
- MOBKL1A|MATS2|MOB4A
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 01
- Reactivity:
- Human
- Host:
- Mouse
- Isotype:
- IgG2a
- ImmunoFluorescence:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient