Anti-Vsir Rabbit Polyclonal Antibody
Produced in rabbits immunized with purified, recombinant Human VISTA/GI24/B7-H5 (rh VISTA/GI24/B7-H5; Catalog#13482-H08H; AAH20568.1; Met1-Ala194). VISTA/GI24/B7-H5 specific IgG was purified by Human VISTA/GI24/B7-H5 affinity chromatography.
Type: Primary
Antigen: VISTA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
- Catalog No:
- 103610-500
- Antigen Symbol:
- Vsir
- Antigen Name:
- V-set immunoregulatory receptor
- Antigen Synonyms:
- B7-H5|C10orf54|GI24|PP2135|DD1alpha|SISP1|B7H5|VISTA|PD-1H
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient