Anti-SCG2 Rabbit Polyclonal Antibody
Produced in rabbits immunized with purified, recombinant Human SCG2 / Secretogranin II (rh SCG2 / Secretogranin II; Catalog#13441-H08H; AAH22509.1; Met1-Met617). Total IgG was purified by Protein A affinity chromatography.
Type: Primary
Antigen: SCG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
- Catalog No:
- 103651-064
- Antigen Symbol:
- SCG2
- Antigen Name:
- Secretogranin II
- Antigen Synonyms:
- EM66|SgII|SN|CHGC
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient