Anti-TSC22D1 Rabbit Monoclonal Antibody [clone:040]
Supplier: Sino Biological
Certificates
About this item
This antibody was obtained from a rabbit immunized with purified, recombinant Human TSC22D1 (rh TSC22D1; Catalog#12724-H07E; Q15714-2; Met1-Ala144).
Type: Primary
Antigen: TSC22D1
Clonality: Monoclonal
Clone: 040
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Specifications
- Catalog No:
- 103656-658
- Antigen Symbol:
- TSC22D1
- Antigen Name:
- TSC22 domain family member 1
- Antigen Synonyms:
- TSC22|TGFB1I4|Ptg-2
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 040
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ImmunoFluorescence:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient