Anti-SCGN Rabbit Monoclonal Antibody [clone:185]
Supplier: Sino Biological
Certificates
About this item
This antibody was obtained from a rabbit immunized with purified, recombinant Human SCGN / Secretagogin (rh SCGN / Secretagogin; Catalog#12411-H07E; NP_008929.2; Asp2-Pro276).
Type: Primary
Antigen: SCGN
Clonality: Monoclonal
Clone: 185
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Specifications
- Catalog No:
- 103636-942
- Antigen Symbol:
- SCGN
- Antigen Name:
- Secretagogin, EF-hand calcium binding protein
- Antigen Synonyms:
- DJ501N12.8|SEGN|SECRET|setagin|CALBL
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 185
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient