Anti-CASP14 Rabbit Monoclonal Antibody [clone:101]
Supplier: Sino Biological
Certificates
About this item
This antibody was obtained from a rabbit immunized with purified, recombinant Human Caspase-14 / CASP14 (rh Caspase-14 / CASP14; Catalog#11856-H07E; NP_036246.1; Ser2-Gln242).
Type: Primary
Antigen: CASP14
Clonality: Monoclonal
Clone: 101
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Specifications
- Catalog No:
- 103635-848
- Antigen Symbol:
- CASP14
- Antigen Name:
- Caspase 14
- Antigen Synonyms:
- ARCI12
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 101
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ImmunoChemistry:
- Yes
- ImmunoFluorescence:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient