Anti-F13B Rabbit Polyclonal Antibody
Produced in rabbits immunized with purified, recombinant Human Coagulation Factor XIII B chain (rh Coagulation Factor XIII B chain; Catalog#11510-H08H; P05160; Met1-Thr661). Coagulation Factor XIII B chain specific IgG was purified by Human Coagulation Factor XIII B chain affinity chromatography.
Type: Primary
Antigen: Factor XIII
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
- Catalog No:
- 103645-094
- Antigen Symbol:
- F13B
- Antigen Name:
- Coagulation factor XIII B chain
- Antigen Synonyms:
- Coagulation factor XIII|Coagulation factor 13|FXIIIB
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient