Anti-HA Mouse Monoclonal Antibody [clone: 3D6B3F5]
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza A virus H3N2 Hemagglutinin. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Type: Primary
Antigen: Hemagglutinin
Clonality: Monoclonal
Clone: 3D6B3F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: H3N2 (A/Brisbane/10/2007) HA
- Catalog No:
- 103620-398
- Antigen Symbol:
- HA
- Antigen Name:
- Hemagglutinin
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 3D6B3F5
- Reactivity:
- Influenza A
- Host:
- Mouse
- Isotype:
- IgG1
- Western Blot:
- Yes
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Frequently Bought Together