Anti-TFRC Rabbit Monoclonal Antibody [clone:040] PerCP (Peridinin-Chlorophyll Protein Complex)
This antibody was obtained from a rabbit immunized with purified, recombinant Human Transferrin Receptor / TFRC / CD71 (rh Transferrin Receptor / TFRC / CD71; Catalog#11020-H07H; AAA61153.1; Cys89-Phe760) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Type: Primary
Antigen: TFRC
Clonality: Monoclonal
Clone: Clone: 040
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
- Catalog No:
- 103631-474
- 103629-362
- Antigen Symbol:
- TFRC
- TFRC
- Antigen Name:
- Transferrin receptor
- Transferrin receptor
- Antigen Synonyms:
- TRFR|T9|TFR1|CD71|p90|TFR|TR
- TRFR|T9|TFR1|CD71|p90|TFR|TR
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PerCP (Peridinin-Chlorophyll Protein Complex)
- PerCP (Peridinin-Chlorophyll Protein Complex)
- Clone:
- 040
- 040
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Flow Cytometry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- Shipping Temperature:
- Ambient
- Ambient