Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-TFRC Rabbit Monoclonal Antibody [clone:040]
Anti-TFRC Rabbit Monoclonal Antibody [clone:040]
Supplier:  Sino Biological
Certificates
undefined
Anti-TFRC Rabbit Monoclonal Antibody [clone:040]
Supplier:  Sino Biological

Specifications

  • Catalog No:
  • 103618-926
  • Antigen Symbol:
  • TFRC
  • Antigen Name:
  • Transferrin receptor
  • Antigen Synonyms:
  • TRFR|T9|TFR1|CD71|p90|TFR|TR
  • Antibody Type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • 040
  • Reactivity:
  • Human
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • ImmunoFluorescence:
  • Yes
  • Flow Cytometry:
  • Yes
  • Form:
  • Liquid
  • Storage Buffer:
  • 0.2 μm filtered solution in PBS
  • Shipping Temperature:
  • Ambient

Specifications

Related Information