Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-IL13Ra1 Rabbit Monoclonal Antibody [clone: 038] (PE (Phycoerythrin))
Anti-IL13Ra1 Rabbit Monoclonal Antibody [clone: 038] (PE (Phycoerythrin))
Supplier:  Sino Biological
Certificates
undefined
Anti-IL13Ra1 Rabbit Monoclonal Antibody [clone: 038] (PE (Phycoerythrin))
Supplier:  Sino Biological

Specifications

  • Catalog No:
  • 103631-018
  • 103629-298
  • Antigen Symbol:
  • IL13Ra1
  • IL13Ra1
  • Antigen Name:
  • Interleukin 13 receptor subunit alpha 1
  • Interleukin 13 receptor subunit alpha 1
  • Antigen Synonyms:
  • CD213A1|NR4|IL-13Ra
  • CD213A1|NR4|IL-13Ra
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • 038
  • 038
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Flow Cytometry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Storage Buffer:
  • PBS solution containing 0.5% BSA and 0.09% sodium azide
  • PBS solution containing 0.5% BSA and 0.09% sodium azide
  • Shipping Temperature:
  • Ambient
  • Ambient

Specifications