Anti-IL13Ra1 Rabbit Monoclonal Antibody [clone: 038] (APC (Allophycocyanin))
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL13RA1 / IL-13RA1 (rh IL13RA1 / IL-13RA1; Catalog#10943-H08H; NP_001551.1; Met1-Thr343) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Type: Primary
Antigen: IL13RA1
Clonality: Monoclonal
Clone: Clone: 038
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
- Catalog No:
- 103631-016
- 103629-296
- Antigen Symbol:
- IL13Ra1
- IL13Ra1
- Antigen Name:
- Interleukin 13 receptor subunit alpha 1
- Interleukin 13 receptor subunit alpha 1
- Antigen Synonyms:
- CD213A1|NR4|IL-13Ra
- CD213A1|NR4|IL-13Ra
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- 038
- 038
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Flow Cytometry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- Shipping Temperature:
- Ambient
- Ambient