Anti-SPARC Rabbit Polyclonal Antibody
Produced in rabbits immunized with purified, recombinant Human SPARC (rh SPARC; Catalog#10929-H08H; NP_003109.1; Met 1-Ile 303). SPARC specific IgG was purified by human SPARC affinity chromatography.
Type: Primary
Antigen: SPARC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
- Catalog No:
- 103618-410
- Antigen Symbol:
- SPARC
- Antigen Name:
- Secreted protein acidic and cysteine rich
- Antigen Synonyms:
- ON|SPARC|BM-40
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Frequently Bought Together