Anti-SPARC Mouse Monoclonal Antibody [clone: 14]
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Osteonectin / SPARC (rh Osteonectin / SPARC; Catalog#10929-H08H; NP_003109.1; Met1-Ile303). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Type: Primary
Antigen: SPARC
Clonality: Monoclonal
Clone: 14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
- Catalog No:
- 103618-398
- Antigen Symbol:
- SPARC
- Antigen Name:
- Secreted protein acidic and cysteine rich
- Antigen Synonyms:
- ON|SPARC|BM-40
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 14
- Reactivity:
- Human
- Host:
- Mouse
- Isotype:
- IgG1
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Frequently Bought Together