Anti-IFNB1 Rabbit Monoclonal Antibody [clone: 040]
Supplier: Sino Biological
Certificates
About this item
This antibody was obtained from a rabbit immunized with purified, recombinant Human IFN beta (rh IFN beta; Catalog#10704-H02H; NP_002167.1; Met1-Asn187).
Type: Primary
Antigen: IFNB
Clonality: Monoclonal
Clone: 040
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Specifications
- Catalog No:
- 103617-186
- Antigen Symbol:
- IFNB1
- Antigen Name:
- Interferon beta 1
- Antigen Synonyms:
- Interferon beta|IFNB|IFB|IFF|IFN-beta
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 040
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient