Anti-TREM1 Mouse Monoclonal Antibody [clone: 07]
Supplier: Sino Biological
Certificates
About this item
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TREM-1 / TREM1 (rh TREM-1 / TREM1; Catalog#10511-H08H; NP_061113.1; Met1-Arg200). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Type: Primary
Antigen: TREM-1
Clonality: Monoclonal
Clone: 07
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- 103616-650
- Antigen Symbol:
- TREM1
- Antigen Name:
- Triggering receptor expressed on myeloid cells 1
- Antigen Synonyms:
- CD354|TREM-1
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 07
- Reactivity:
- Human
- Host:
- Mouse
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient