Anti-FCAR Mouse Monoclonal Antibody [clone: 02] (APC (Allophycocyanin))
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD89 / FCAR (rh CD89 / FCAR; Catalog#10414-H08H; NP_001991.1; Met 1-Asn 227) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Type: Primary
Antigen: CD89
Clonality: Monoclonal
Clone: Clone: 02
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
- Catalog No:
- 103629-034
- 103630-764
- Antigen Symbol:
- FCAR
- FCAR
- Antigen Name:
- Fc alpha receptor
- Fc alpha receptor
- Antigen Synonyms:
- FcalphaRI|CTB-61M7.2|CD89|FCAR|XXbac-BPG230H20.5
- FcalphaRI|CTB-61M7.2|CD89|FCAR|XXbac-BPG230H20.5
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- 02
- 02
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Flow Cytometry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- Shipping Temperature:
- Ambient
- Ambient