Anti-IL4R Rabbit Monoclonal Antibody [clone: 102]
Supplier: Sino Biological
Certificates
About this item
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL4R (rh IL4R; Catalog#10402-H08H; NP_000409.1; Met 1-His 232).
Type: Primary
Antigen: IL4R
Clonality: Monoclonal
Clone: 102
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Specifications
- Catalog No:
- 103649-114
- Antigen Symbol:
- IL4R
- Antigen Name:
- Interleukin 4 receptor
- Antigen Synonyms:
- IL4R|IL-4RA|IL4RA|CD124
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 102
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Specifications
Frequently Bought Together