Anti-CD4 Mouse Monoclonal Antibody [clone: 08] (APC (Allophycocyanin))
Supplier: Sino Biological
Certificates
About this item
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD4 (rh CD4; Catalog#10400-H08H; NP_000607.1; Met1-Trp390) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Type: Primary
Antigen: Anti-CD4 Antibody
Clonality: Monoclonal
Clone: Clone: 08
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Specifications
- Catalog No:
- 103629-016
- 103630-738
- Antigen Symbol:
- CD4
- CD4
- Antigen Name:
- CD4 molecule
- CD4 molecule
- Antigen Synonyms:
- CD4mut
- CD4mut
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- 08
- 08
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Flow Cytometry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- Shipping Temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together