Anti-CD4 Mouse Monoclonal Antibody [clone: 6F7B4C5]
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD4 (rh CD4; Catalog#10400-H08H; NP_000607.1; Met 1-Trp 390). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Type: Primary
Antigen: Anti-CD4 Antibody
Clonality: Monoclonal
Clone: 6F7B4C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
- Catalog No:
- 103649-068
- Antigen Symbol:
- CD4
- Antigen Name:
- CD4 molecule
- Antigen Synonyms:
- CD4mut
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 6F7B4C5
- Reactivity:
- Human
- Host:
- Mouse
- Isotype:
- IgG1
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Frequently Bought Together