Anti-ICAM1 Mouse Monoclonal Antibody [clone: 01] (PerCP (Peridinin-Chlorophyll Protein Complex))
Supplier: Sino Biological
Certificates
About this item
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ICAM-1 / CD54 (rh ICAM-1 / CD54; Catalog#10346-H08H; NP_000192.2; Met 1-Glu 480) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Type: Primary
Antigen: ICAM-1 (PerCP)
Clonality: Monoclonal
Clone: Clone: 01
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Specifications
- Catalog No:
- 103628-964
- 103630-690
- Antigen Symbol:
- ICAM1
- ICAM1
- Antigen Name:
- Intercellular adhesion molecule 1
- Intercellular adhesion molecule 1
- Antigen Synonyms:
- ICAM-1|BB2|CD54|P3.58
- ICAM-1|BB2|CD54|P3.58
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PerCP (Peridinin-Chlorophyll Protein Complex)
- PerCP (Peridinin-Chlorophyll Protein Complex)
- Clone:
- 01
- 01
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Flow Cytometry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- Shipping Temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together