Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-ICAM1 Mouse Monoclonal Antibody [clone: 01] (APC (Allophycocyanin))
 
undefined
Anti-ICAM1 Mouse Monoclonal Antibody [clone: 01] (APC (Allophycocyanin))

 

  • Catalog No:
  • 103630-688
  • 103628-962
  • Antigen Symbol:
  • ICAM1
  • ICAM1
  • Antigen Name:
  • Intercellular adhesion molecule 1
  • Intercellular adhesion molecule 1
  • Antigen Synonyms:
  • ICAM-1|BB2|CD54|P3.58
  • ICAM-1|BB2|CD54|P3.58
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • 01
  • 01
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • Flow Cytometry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Storage Buffer:
  • PBS solution containing 0.5% BSA and 0.09% sodium azide
  • PBS solution containing 0.5% BSA and 0.09% sodium azide
  • Shipping Temperature:
  • Ambient
  • Ambient

 

Frequently Bought Together