Anti-PRKAA1 Rabbit Polyclonal Antibody
Supplier: Sino Biological
Certificates
About this item
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human PRKAA1, and purified by antigen affinity chromatography.
Type: Primary
Antigen: PRKAA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 103646-938
- Antigen Symbol:
- PRKAA1
- Antigen Name:
- Protein kinase AMP-activated catalytic subunit alpha 1
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Isotype:
- IgG
- ImmunoChemistry:
- Yes
- ImmunoFluorescence:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient