Anti-ENG Mouse Monoclonal Antibody [clone: 13] (APC (Allophycocyanin))
Supplier: Sino Biological
Certificates
About this item
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Endoglin / CD105 (rh Endoglin / CD105 ; Catalog#10149-H08H; NP_001108225.1; Met1-Gly586) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Type: Primary
Antigen: ENG
Clonality: Monoclonal
Clone: Clone: 13
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b
Reactivity: Human
Specifications
- Catalog No:
- 103631-168
- 103630-510
- Antigen Symbol:
- ENG
- ENG
- Antigen Name:
- Endoglin
- Endoglin
- Antigen Synonyms:
- END|ORW1|HHT1
- END|ORW1|HHT1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- 13
- 13
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2b
- IgG2b
- Flow Cytometry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- PBS solution containing 0.5% BSA and 0.09% sodium azide
- Shipping Temperature:
- Ambient
- Ambient