Anti-RPS3 Rabbit Polyclonal Antibody
Supplier: Sino Biological
Certificates
About this item
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human RPS3, and purified by antigen affinity chromatography.
Type: Primary
Antigen: RPS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 103593-426
- Antigen Symbol:
- RPS3
- Antigen Name:
- Ribosomal protein S3
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Isotype:
- IgG
- Western Blot:
- Yes
- ImmunoPrecipitation:
- Yes
- ImmunoFluorescence:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient