Anti-IL1A Rabbit Polyclonal Antibody
Supplier: Sino Biological
Certificates
About this item
Produced in rabbits immunized with purified, recombinant Human IL-1 alpha / IL-1F1 / IL1A (rh IL-1 alpha / IL-1F1 / IL1A; Catalog#10128-HNCH; NP_000566.3; Ser113-Ala271). IL-1 alpha / IL-1F1 / IL1A specific IgG was purified by Human IL-1 alpha / IL-1F1 / IL1A affinity chromatography.
Type: Primary
Antigen: IL-1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Specifications
- Catalog No:
- 103639-728
- Antigen Symbol:
- IL1A
- Antigen Name:
- Interleukin 1 alpha
- Antigen Synonyms:
- IL-1F1|IL1F1|IL-1A|IL1|IL1-ALPHA
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ELISA:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
Specifications
Frequently Bought Together