Anti-THY1 Rabbit Polyclonal Antibody
Supplier: Sino Biological
About this item
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the human CD90, and purified by antigen affinity chromatography.
Type: Primary
Antigen: Thy1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
1 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the human CD90, and purified by antigen affinity chromatography.
Type: Primary
Antigen: Thy1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Documents
Specifications for Product Family
Specifications
- Catalog No:
- 103638-860
- 103591-164
- 103591-166
- Antigen Symbol:
- THY1
- THY1
- THY1
- Antigen Name:
- Thy-1 cell surface antigen
- Thy-1 cell surface antigen
- Thy-1 cell surface antigen
- Antigen Synonyms:
- CD90
- CD90
- CD90
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- 0.2 μm filtered solution in PBS
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient
- Ambient
- Ambient
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- 50 µlDiscontinued-Supplier Number: -Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.103638-860Antigen SymbolTHY1Antigen NameThy-1 cell surface antigenAntigen SynonymsCD90Antibody TypePrimaryClonalityPolyclonalConjugationUnconjugatedReactivityHumanHostRabbitIsotypeIgGWestern BlotYesFormLiquidStorage Buffer0.2 μm filtered solution in PBSShipping TemperatureAmbient - Supplier Number: 100673-T32-100Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.103591-164Antigen SymbolTHY1Antigen NameThy-1 cell surface antigenAntigen SynonymsCD90Antibody TypePrimaryClonalityPolyclonalConjugationUnconjugatedReactivityHumanHostRabbitIsotypeIgGWestern BlotYesFormLiquidStorage Buffer0.2 μm filtered solution in PBSShipping TemperatureAmbient - 200 µlDiscontinued-Supplier Number: -Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.103591-166Antigen SymbolTHY1Antigen NameThy-1 cell surface antigenAntigen SynonymsCD90Antibody TypePrimaryClonalityPolyclonalConjugationUnconjugatedReactivityHumanHostRabbitIsotypeIgGWestern BlotYesFormLiquidStorage Buffer0.2 μm filtered solution in PBSShipping TemperatureAmbient



