Anti-NFIB Rabbit Polyclonal Antibody
Produced in rabbits immunized with A synthetic peptide corresponding to the C-terminus of the human NF1B, and purified by antigen affinity chromatography.
Type: Primary
Antigen: NF1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human;Mouse
- Catalog No:
- 103592-376
- Antigen Symbol:
- NFIB
- Antigen Name:
- Nuclear factor I B
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Mouse
- Host:
- Rabbit
- Isotype:
- IgG
- Western Blot:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm filtered solution in PBS
- Shipping Temperature:
- Ambient