ContactUsLinkComponent
Swivel Clamp Holder, United Scientific Supplies
Swivel Clamp Holder, United Scientific Supplies
Catalog # 76293-418
undefined
Swivel Clamp Holder, United Scientific Supplies
Catalog # 76293-418
Supplier Number:  CLMS01-VWR
Specifications
  • Description:
    Swivel Clamp Holder
  • Cat. No.:
    76293-418
Specifications
About this item
Plated steel double clamp.
  • Rotates 360 degrees
  • Plated steel construction
Accepts rods up to 3/4" diameter (19 mm). One clamp can be rotated 360° along its axis and locked in any position. Length across clamps is 4.5".