Static Dissipative Nylon Bristle Applicator Brush, Gordon Brush
76254-224
- Bristle material:Nylon
- Handle material:Stainless Steel
- Description:Static Dissipative Nylon Bristle Applicator Brush
- Brush head width:5/16"
- Cat. No.:76254-224
- Handle length:41/2"
- Trim length:⁵⁄₁₆"
Nylon bristle applicator brush with stainless steel handle.
- anti-static, static dissipative
- Lightweight, chemical resistant, non-magnetic, stainless steel handle
- Adhesive-free to prevent contamination on surface areas caused by leaching
- Maximum use temperature of 225°F
Manufactured using strong mechanical clamping for maximum fiber retention.