Supreme Pliers, JH Williams Tool Group
76238-882
- Description:Supreme Plier, Chain Nose, Smooth
- Environmentally Preferable:
- Cat. No.:76238-882
Excellent pliers for general electronics work and precise wire bending.
Supreme series pliers features a precision screw and nut in an advanced lap joint design. Joint play is held to a minimum, ensuring extremely precise alignment of the jaws even at the extreme tips.