ContactUsLinkComponent
Self-Wringing Sponge Mop Head, Foamtec
Self-Wringing Sponge Mop Head, Foamtec
  76246-632
Self-Wringing Sponge Mop Head, Foamtec
  76246-632
 :  FS601-12
  • For Use With:
    14" FT401 handles
  • Description:
    Self-Wringing Sponge Mop Head
  • Cat. No.:
    76246-632
UltraMOP SW Cleanroom sponge mop head with a galvanized steel bracket is designed to fit on all FHP style 14″ self-wringing handles.
The 14″ sponge mop head is constructed of a tear-resistant, open cell polyurethane sponge that cleans, absorbs spills and applies even volumes of disinfectants.