PureFlo® MZP Filter Series Capsule, Zenpure Americas
Catalog # 76250-032
Supplier: ZENPURE AMERICAS C/O ST GOBAIN TE
Specifications
- Micron Rating:0.2 µ
- Description:PureFlo® MZP Filter Series Capsule
- Environmentally Preferable:
- Cat. No.:76250-032
- Grade:Pharma
- Inlet/outlet connections:1/4" Hose Barb Connections
Specifications
About this item
Featured with gamma stable polyproylene shell.