Reusable Altessa Grid Cleanroom Coveralls, Uniform Technology
Cleanroom coveralls are manufactured from 99% polyester/1% carbon fabric.
- Military collar
- Raglan sleeves
- ESD knit wrists
- Coil zipper with top snaps
- Snap closures at ankle and back of legs
Suitable for ISO Class 5 or higher cleanrooms.
- Catalog No:
- 76515-672
- 76515-670
- 76515-668
- 76515-666
- 76515-676
- 76515-674
- 76515-656
- 76515-654
- 76515-652
- 76515-650
- 76515-664
- 76515-662
- 76515-660
- 76515-658
- 76515-680
- 76515-678
- 76515-648
- 76515-646
- 76515-644
- 76515-642
- 76515-682
- Model:
- CC1245-74
- CC1245-74
- CC1245-74
- CC1245-74
- CC1245-74
- CC1245-74
- CC1245-74
- CC1245-74
- CC1245-74
- CC1245-74
- CC1245-74
- CC1245-74
- CC1245-74
- CC1245-74
- CC1245-74
- CC1245-74
- CC1245-74
- CC1245-74
- CC1245-74
- CC1245-74
- CC1245-74
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- Fabric:
- 99% polyester/1% carbon fabric
- 99% polyester/1% carbon fabric
- 99% polyester/1% carbon fabric
- 99% polyester/1% carbon fabric
- 99% polyester/1% carbon fabric
- 99% polyester/1% carbon fabric
- 99% polyester/1% carbon fabric
- 99% polyester/1% carbon fabric
- 99% polyester/1% carbon fabric
- 99% polyester/1% carbon fabric
- 99% polyester/1% carbon fabric
- 99% polyester/1% carbon fabric
- 99% polyester/1% carbon fabric
- 99% polyester/1% carbon fabric
- 99% polyester/1% carbon fabric
- 99% polyester/1% carbon fabric
- 99% polyester/1% carbon fabric
- 99% polyester/1% carbon fabric
- 99% polyester/1% carbon fabric
- 99% polyester/1% carbon fabric
- 99% polyester/1% carbon fabric