Order Entry
Puerto Rico
ContactUsLinkComponent
HiTrap Columns, MabSelect PrismA, Cytiva
HiTrap Columns, MabSelect PrismA, Cytiva
Supplier:  Cytiva
Certificates
undefined
HiTrap Columns, MabSelect PrismA, Cytiva
Supplier:  Cytiva
Certificates

Specifications

  • Catalog No:
  • 76237-730
  • 76237-728
  • 76235-124
  • 76237-726
  • Binding Capacity:
  • ~ 40 mg polyclonal IgG/ml resin, 2 min residence time
    ~ 80 mg polyclonal IgG/ml resin, 6 min residence time
  • ~ 40 mg polyclonal IgG/ml resin, 2 min residence time
    ~ 80 mg polyclonal IgG/ml resin, 6 min residence time
  • ~ 40 mg polyclonal IgG/ml resin, 2 min residence time
    ~ 80 mg polyclonal IgG/ml resin, 6 min residence time
  • ~ 40 mg polyclonal IgG/ml resin, 2 min residence time
    ~ 80 mg polyclonal IgG/ml resin, 6 min residence time
  • Max. Pressure:
  • 5 bar
  • 5 bar
  • 5 bar
  • 5 bar
  • pH Range:
  • 3 - 12
  • 3 - 12
  • 3 - 12
  • 3 - 12
  • Temperature Limit:
  • 40 °C
  • 40 °C
  • 40 °C
  • 40 °C
  • Storage:
  • 20% ethanol, 2...8 °C
  • 20% ethanol, 2...8 °C
  • 20% ethanol, 2...8 °C
  • 20% ethanol, 2...8 °C

Specifications