HardyVal™ Tryptic Soy Broth Ampoule, USP, with Red Dye
Catalog # 76018-782
Supplier: Hardy Diagnostics
CAS Number:
Specifications
- Pack type:Ampoule
- Description:Tryptic Soy Broth, USP, with red dye
- Size:5 ml
- Cat. No.:76018-782
- Fill volume:3 ml
Specifications
About this item
Compliance made easy to verify the efficacy of aseptic technique.
- Non-toxic red dye is added to the TSB to easily visualize the sample being transferred
This 3 ml glass ampoules of media are designed to complement personal aseptic technique proficiency tests as described in USP procedures.