Order Entry
Puerto Rico
ContactUsLinkComponent
HardyVal™ Tryptic Soy Broth, USP, Media-Bag™, Hardy Diagnostics
HardyVal™ Tryptic Soy Broth, USP, Media-Bag™, Hardy Diagnostics
Catalog # 75814-002
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
HardyVal™ Tryptic Soy Broth, USP, Media-Bag™, Hardy Diagnostics
Catalog # 75814-002
Supplier:  Hardy Diagnostics
Supplier Number:  U100
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Pack type:
    Bag, double zippered, 2 ml
  • Description:
    Tryptic Soy Broth (TSB), USP
  • Size:
    10/pack
  • Cat. No.:
    75814-002
  • Volume:
    100 ml
  • Supplier no.:
    U100

Specifications

About this item

Compliance made easy to verify the efficacy of aseptic technique.

  • The Media-Bag™ has both a spike-port and needle-port
  • Terminally autoclaved for high sterility assurance
  • Hang Notch on the top of the Media Bag™ for convenience
  • Packaged in double 2 ml zippered bags
  • Growth promotion testing performed on each lot of media

Tryptic Soy Broth is recommended for use as a general-purpose medium for the isolation and cultivation of a wide variety of bacteria and fungi. The U.S. Pharmacopeia National Formulary (USP) describes its use for sterility testing.

Ideal for verifying aseptic technique of pharmacists, nurses, or pharmaceutical manufacturing technicians.

Quality tested to USP 71 standards, manufactured in an ISO 13485 facility.