HardyVal™ CSP Medium Complexity Kit, Basic, Hardy Diagnostics
Product Details & Documents
Compliance made easy to verify the efficacy of aseptic technique.
- The product is quality control tested for growth promotion, pH, and sterility
The Medium Complexity Level Kit, Basic is recommended for simulating medium risk level compounding manipulations and verification of aseptic transfer techniques of multiple solutions. Kit performs one complete aseptic technique verification challenge test for one technician.
Delivery information: Each Kit Contains:
Tryptic Soy Broth (TSB), 50 ml Needle Port Septum Serum Vial (3 vials)
Sterile 50 ml Serum Vials, empty (6 vials)
Sterile 20 ml Serum Vials, empty (3 vials)
Whirl-Pak™ Bag (1 bag)
Results Log Sheet (1 sheet)
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Description:HardyVal™ CSP Medium Complexity Kit, Basic
- Cat. No.:89408-158
- Storage Temperature:–2-30°C
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



