ContactUsLinkComponent
Wickerham Card, Hardy Diagnostics
Wickerham Card, Hardy Diagnostics
Catalog # 76288-330
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Wickerham Card, Hardy Diagnostics
Catalog # 76288-330
Supplier:  Hardy Diagnostics
Supplier Number:  Z08
Product available on GSA Advantage®
CAS Number:  
Specifications
  • Description:
    Wickerham Card
  • Size:
    2×3"
  • Cat. No.:
    76288-330
Specifications
About this item
Designed for turbidity adjustments to a McFarland standard.
A plastic laminated card with thick black and white lines to facilitate the preparation of bacterial and yeast suspensions when comparing the turbidity to a McFarland Standard. A handy visual aid to be used for the preparation of inocula for the disk diffusion and other tests that require a specified McFarland turbidity.