Potato Flake Agar, Hardy Diagnostics
Hardy Diagnostics Potato Flake Media is used in the cultivation, isolation, and induction of sporulation of fungi from clinical specimens.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.