Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant IL-28B/IFN-lambda-3 Protein (His Tag) (from HEK293 Cells)
Mouse Recombinant IL-28B/IFN-lambda-3 Protein (His Tag) (from HEK293 Cells)
Supplier:  Sino Biological
Certificates
undefined
Mouse Recombinant IL-28B/IFN-lambda-3 Protein (His Tag) (from HEK293 Cells)
Supplier:  Sino Biological

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg protein as determined by the LAL method.
  • Biological Activity:
    Measured in antiviral assay using HepG2 cells infected with vesicular stomatitis virus.
    The ED50 for this effect is typically 44.6-178.4 ng/ml.
  • Gene ID:
    NP_796370.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    INF-alpha|IFL-1|IL-28B|INF-lambda|Il28b|Il28
  • Protein/Peptide Name:
    IFNL3
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant mouse Ifnl3 consists 185 amino acids and predicts a molecular mass of 21.1 kDa.
  • Sequence:
    Met1-Val193
  • Shipping Temperature:
    Ambient

Specifications

Related Information

Frequently Bought Together