Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant CD45/PTPRC Protein (aa 453-1152) (from Baculovirus Insect Cells)
Mouse Recombinant CD45/PTPRC Protein (aa 453-1152) (from Baculovirus Insect Cells)
Supplier:  Sino Biological
Certificates
undefined
Mouse Recombinant CD45/PTPRC Protein (aa 453-1152) (from Baculovirus Insect Cells)
Supplier:  Sino Biological

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Baculovirus (Insect cells)
  • Species:
    Mouse
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    1. The specific activity was determined to be 9168 nmol/min/mg using p-nitrophenyl phosphate as substrate.
    2. Measured by its binding ability in a functional ELISA. Immobilized mouse PTPRC (453-1152) at 10 μg/ml (100 μl/well) can bind biotinylated human Galectin-1 (Cat:10290-HNAE-E) with a linear range of 0.31-2.5 μg/ml.
  • Gene ID:
    AAA39458.1
  • Reconstitution Instructions:
    A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    CD45R|B220|CD45|GP180|T200|L-CA|LY5|LCA
  • Protein/Peptide Name:
    PTPRC
  • Purity:
    >85% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant mouse PTPRC consists of 706 amino acids and has a calculated molecular mass of 81 kDa. The recombinant protein migrates as an approximately 93 kDa band in SDS-PAGE under reducing conditions.
  • Sequence:
    Arg453-Ser1152
  • Shipping Temperature:
    Ambient

Specifications

Related Information