Order Entry
Puerto Rico
ContactUsLinkComponent
 
Mouse Recombinant REN1/Renin-1 Protein (His Tag) (from HEK293 Cells)
 
undefined
Mouse Recombinant REN1/Renin-1 Protein (His Tag) (from HEK293 Cells)

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    1. Measured by its ability to cleave the fluorogenic peptide substrate 5­FAM/QXL™ 520 (Peti­Peterdi, J. et al., 2009, Physiology 24:88.). The specific activity is >20 pmoles/min/μg.
    2. Immobilized mouse REN1-His (Cat: 50279-M08H) at 10 µg/ml (100 μl/well) can bind biotinylated human AGT-His (Cat: 10994-H08H) with a linear range of 31.25 - 250 ng/ml.
  • Gene ID:
    NP_112469.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    Ren1c|Ren-A|Ren1d|Rnr|Ren-2|D19352|Ren-1|Rn-1|Ren2|Rn-2|Ren-B|Ren|Ren1
  • Protein/Peptide Name:
    REN
  • Purity:
    >97% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant mouse REN1 consists of 392 amino acids and has a predicted molecular mass of 43.2 kDa. As a result of glycosylation, the apparent molecular mass of rmREN1 is approximately 48-55 kDa in SDS-PAGE under reducing conditions.
  • Sequence:
    Met1-Arg402
  • Shipping Temperature:
    Ambient

 

Frequently Bought Together