Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse/Rat Recombinant aFGF/FGF1 Protein (from E. coli)
Mouse/Rat Recombinant aFGF/FGF1 Protein (from E. coli)
Supplier:  Sino Biological
Certificates
undefined
Mouse/Rat Recombinant aFGF/FGF1 Protein (from E. coli)
Supplier:  Sino Biological

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
    Mouse
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    Please contact us for more information.
  • Biological Activity:
    Measured in a cell proliferation assay using Balb/c 3T3 mouse embryonic fibroblast cells. The ED50 for this effect is 0.04 - 0.2 ng/ml.
  • Gene ID:
    NP_034327.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    AFGF|ECGFA|ECGF-beta|HBGF-1|FGF-alpha|HBGF1|FGFA|GLIO703|FGF-1|ECGF|ECGFB
  • Protein/Peptide Name:
    FGF1
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant mouse / rat FGF1 consisting of 141 amino acids and has a calculated molecular mass of 16 kDa as estimated in SDS-PAGE under reducing conditions.
  • Sequence:
    Phe16-Asp155
  • Shipping Temperature:
    Ambient

Specifications

Related Information